Learn More
Grainger EOL Resistor PK7
Catalog No. 502765213
Encompass_Preferred
The Edwards Signaling EOL Resistor is an end-of-line resistor used for fire alarm systems. It has a resistance of 4.7k ohms and a power rating of 1/4 watt. The resistor has axial leads and measures 0.4" L x 0.2" W. It can be used to provide supervision of Class B wiring in fire alarm devices. The EOL resistor provides an end-of-line indication and is often wired in series with the last device on the circuit. This Edwards Signaling component allows the fire panel to monitor the circuit for faults.
Catalog No. 50-276-5213 Supplier Grainger Supplier No. 16X397
The Edwards Signaling EOL Resistor is an end-of-line resistor used for fire alarm systems. It has a resistance of 4.7k ohms and a power rating of 1/4 watt. The resistor has axial leads and measures 0.4" L x 0.2" W. It can be used to provide supervision of Class B wiring in fire alarm devices. The EOL resistor provides an end-of-line indication and is often wired in series with the last device on the circuit. This Edwards Signaling component allows the fire panel to monitor the circuit for faults.