Learn More
Grainger EOL Resistor
Catalog No. 502766958
Encompass_Preferred
The Edwards Signaling EOL Resistor is an end-of-line resistor used for fire alarm systems. It has a resistance value of 4.7k ohms and a power rating of 1/4 watt. The resistor has axial leads and measures 0.4" L x 0.2" W. It is RoHS compliant and is made of flame retardant material. The EOL resistor provides supervision of wiring in alarm circuits and signals the panel if the circuit is open. It is compatible with fire panels from Edwards, GE, and EST. The EOL resistor comes in a package of 10.
Catalog No. 50-276-6958 Supplier Grainger Supplier No. 16X395
The Edwards Signaling EOL Resistor is an end-of-line resistor used for fire alarm systems. It has a resistance value of 4.7k ohms and a power rating of 1/4 watt. The resistor has axial leads and measures 0.4" L x 0.2" W. It is RoHS compliant and is made of flame retardant material. The EOL resistor provides supervision of wiring in alarm circuits and signals the panel if the circuit is open. It is compatible with fire panels from Edwards, GE, and EST. The EOL resistor comes in a package of 10.