Learn More
Aviva Systems Biology Corp EOMES Antibody - middle region (ARP38626_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38626_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EOMES. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EOMES. Alternate names: TBR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56733