Learn More
Aviva Systems Biology Corp Eomes Antibody - middle region (ARP37879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eomes. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C77258, Tbr2, TBR-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-698