Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology EOMES Monoclonal Antibody, Rabbit IgG, Unconjugated, 100 μl
SDP

Catalog No. 502731020
Encompass

EOMES (D8D1R) Rabbit Monoclonal Antibody recognizes endogenous levels of total EOMES protein. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro180 of the human EOMES protein. EOMES, also known as Tbr2, is a master regulator of mesoderm formation and is essential for neurogenesis, trophoblast formation, and the differentiation of specific T cell subsets.

  • Validated for Western blotting, immunofluorescence, flow cytometry, chromatin IP, and CUT&RUN applications.
  • Host species: rabbit.
  • Reactivity: human.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Molecular weight: 75 kDa and 85 kDa.

Catalog No. 50-273-1020 Supplier Cell Signaling Technology Supplier No. 81493S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EOMES (D8D1R) Rabbit Monoclonal Antibody recognizes endogenous levels of total EOMES protein. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro180 of the human EOMES protein. EOMES, also known as Tbr2, is a master regulator of mesoderm formation and is essential for neurogenesis, trophoblast formation, and the differentiation of specific T cell subsets.

  • Validated for Western blotting, immunofluorescence, flow cytometry, chromatin IP, and CUT&RUN applications.
  • Host species: rabbit.
  • Reactivity: human.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Molecular weight: 75 kDa and 85 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.