Learn More
Cayman Chemical EOPt18 028 018 2-d11 100UG
SDP
Supplier:  Cayman Chemical 9004339100 UG
Encompass_Preferred
An internal standard for the quantification of EOP (t18:0/28:0/18:2) by GC- or LC-MS
Catalog No. 50-289-5804