Learn More
Cayman Chemical EOPt18 028 018 2-d11 250UG
SDP
Catalog No. 502895805
Encompass_Preferred
An internal standard for the quantification of EOP (t18:0/28:0/18:2) by GC- or LC-MS
Catalog No. 50-289-5805 Supplier Cayman Chemical Supplier No. 9004339250 UG
An internal standard for the quantification of EOP (t18:0/28:0/18:2) by GC- or LC-MS