Description
- Portable, handheld electrochemical device designed for fast and accurate point-of-care testing of blood ketone and blood glucose levels in cattle
Specifications
Specifications
| Product Type | Blood Glucose Meter Kit |
| Analysis Time | 5 to 10 sec. |
| Sample Size | 0.8 μL |
| Quantity | 1 Ea. |