Learn More
Cayman Chemical EOSd18 132 118 2 100ug
SDP
Catalog No. 502986956
Encompass_Preferred
An essential ceramide for prevention of epidermal water loss; found in the outer epidermis of mammals; consecutive regio- and stereospecific oxygenation of the linoleate portion by 12(R)-LO and eLOX3 is essential for the maintenance of the epidermal barrier to prevent water loss
Catalog No. 50-298-6956 Supplier Cayman Chemical Supplier No. 22442100 UG
An essential ceramide for prevention of epidermal water loss; found in the outer epidermis of mammals; consecutive regio- and stereospecific oxygenation of the linoleate portion by 12(R)-LO and eLOX3 is essential for the maintenance of the epidermal barrier to prevent water loss