Learn More
AdipoGen Eosin-5-isothiocyanate
SDP
Catalog No. 502560996
Encompass_Preferred
Chemical. CAS 60520-47-0. Formula C21H7Br4NO5S. MW 705. Synthetic. Fluorescent probe. Long wavelength red labeling derivative for proteins. Can be used as a reversible inhibitor of the erythrocyte calcium pump as well as an effective phosphorescent probe in measurement of rotational diffusion of proteins and in fluorescence energy transfer FRET studies. EITC tagged molecules may be used together with fluorescein-5-isothiocyanate FITC tagged molecules in fluoresce resonance energy transfer FRET based assays. Spectral properties Abs/Em = 520/544 nm
Catalog No. 50-256-0996 Supplier AdipoGen Supplier No. CDXE0013M050
Chemical. CAS 60520-47-0. Formula C21H7Br4NO5S. MW 705. Synthetic. Fluorescent probe. Long wavelength red labeling derivative for proteins. Can be used as a reversible inhibitor of the erythrocyte calcium pump as well as an effective phosphorescent probe in measurement of rotational diffusion of proteins and in fluorescence energy transfer FRET studies. EITC tagged molecules may be used together with fluorescein-5-isothiocyanate FITC tagged molecules in fluoresce resonance energy transfer FRET based assays. Spectral properties Abs/Em = 520/544 nm