Learn More
Biotium EOSIN-5-Maleimide
Catalog No. 501964840
Encompass_Preferred
Eosin-5-maleimide is a good photosensitizer and can be used to selectively label thiols. It has a quantum yield of 0.57 for singlet oxygen generation, which makes it useful for photoconversion of electron-rich materials such as diaminobenzidine (DAB) or fluorescent dyes into highly electron-dense materials for high resolution electron microscopy studies. Additionally, eosin is a good fluorescence acceptor for fluorescein (FAM), dansyl and coumarin dyes in fluorescence resonance energy transfer (FRET). Properties: Ex/Em (MeOH) = 524/545 nm; e = 100,000; Orange solid soluble in DMF or DMSO; Store at -20°C and protect from light; C24H9Br4NO7; MW: 742.95.
Catalog No. 50-196-4840 Supplier Biotium Supplier No. 92013
Eosin-5-maleimide is a good photosensitizer and can be used to selectively label thiols. It has a quantum yield of 0.57 for singlet oxygen generation, which makes it useful for photoconversion of electron-rich materials such as diaminobenzidine (DAB) or fluorescent dyes into highly electron-dense materials for high resolution electron microscopy studies. Additionally, eosin is a good fluorescence acceptor for fluorescein (FAM), dansyl and coumarin dyes in fluorescence resonance energy transfer (FRET). Properties: Ex/Em (MeOH) = 524/545 nm; e = 100,000; Orange solid soluble in DMF or DMSO; Store at -20°C and protect from light; C24H9Br4NO7; MW: 742.95.