Learn More
Invitrogen™ Eosin-5-Maleimide
Description
Bioconjugates made with the thiol-reactive eosin-5-maleimide can be used as phosphorescent probes or as photosensitizers. With its high quantum yield (∼0.57) for singlet oxygen generation, eosin and its conjugates can be used as effective photooxidizers of diaminobenzidine (DAB) in high-resolution electron microscopy studies and in correlated fluorescence and electron microscopy applications.
Specifications
Specifications
| Quantity | 25 mg |
| Product Type | Maleimide |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Thiol |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Maleimide |
| Label or Dye | Eosin |
| Label Type | Classic Dyes |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.