Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin B, pure, certified

Catalog No. p-3756499
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409420250 25 g Glass bottle
AC409421000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409420250 Supplier Thermo Scientific Chemicals Supplier No. 409420250
Only null left
Add to Cart

CAS: 548-24-3 | C20H6Br2N2Na2O9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
MDL Number MFCD00005041
Synonym 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish

Specifications

Absorption Ratio 0.96 to 1.22
Dye Content 85% min.
Beilstein 19, 233
Merck Index 15, 3657
Infrared Spectrum Authentic
Lambda Max 511 to 520nm (in water + 1% Na2CO3)
Formula Weight 624.06
Grade Pure
Packaging Glass bottle
Color Brown to Purple/Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Eosin B, Pure
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.