Learn More
Abcam Eosin 5-isothiocyanate, 25MG
Supplier:  Abcam AB27430325MG
Encompass_Preferred
MW 705 Da, Purity >95%. Red fluorescent label used to label amino groups on proteins. Can also be used as a reversible inhibitor of the erythrocyte calcium pump. An effective phosphorescent probe in protein rotational studies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-205