Learn More
Cayman Chemical EosIn-Labeled InsulIn 1ea
SDP
Catalog No. 502969469
Encompass_Preferred
A fluorogenic substrate for thioredoxin; eosin is released upon reduction by thioredoxin and its fluorescence can be used to quantify thioredoxin activity; ex/em = 520/545 nm, respectively
Catalog No. 50-296-9469 Supplier Cayman Chemical Supplier No. 154951 EA
A fluorogenic substrate for thioredoxin; eosin is released upon reduction by thioredoxin and its fluorescence can be used to quantify thioredoxin activity; ex/em = 520/545 nm, respectively