Learn More
Bioworld Eosin Methylene Blue Agar (HOLT, Harris), 500 g
Catalog No. 50254301
Encompass_Preferred
Eosin Methylene Blue Agar, Modified (formula of Holt-Harris and Teague)Slightly selective and differential medium for the isolation, cultivation and differentiation of gram-negative enteric bacillipH: 7.2Working Concentration: 36 g/LDehydrated Appearance: light red-purple, homogeneous powderPrepared Appearance: dark blue purple agarSoluble in boiling waterComposition:Pancreatic Digest of Gelatin Lactose Sucrose Dipotassium Phosphate Eosin Y Methylene Blue Agar
Catalog No. 50-254-301 Supplier Bioworld Supplier No. 306200251
Eosin Methylene Blue Agar, Modified (formula of Holt-Harris and Teague)Slightly selective and differential medium for the isolation, cultivation and differentiation of gram-negative enteric bacillipH: 7.2Working Concentration: 36 g/LDehydrated Appearance: light red-purple, homogeneous powderPrepared Appearance: dark blue purple agarSoluble in boiling waterComposition:Pancreatic Digest of Gelatin Lactose Sucrose Dipotassium Phosphate Eosin Y Methylene Blue Agar