Learn More
Carolina™ Eosin Methylene Blue Agar, Prepared Media Plates 100 x 15mm
SDPSCI_ED
Catalog No. S42831 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S42831 10/Pk.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S42831 Supplier Carolina™ Supplier No. 821522
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Carefully tested for sterility and ability to support the characteristic growth of appropriate organisms.
  • Used for characteristic growth of organisms
Specifications
Sterility Sterile
For Use With (Application) Culture of organisms
Quantity 10/Pk.
Diameter (Metric) 100 mm