Learn More
Newcomer Supply Eosin-Phloxine Stain Set. Intended to give an extra fine touch to routine hematoxylin and eosin (H&E) stains and can be used in either manual or automated staining platforms.
SDP
Catalog No. NC0068658
Encompass
Solution A: Eosin Y Stock Stain 1%, Aqueous 1000 ml, Solution B: Phloxine B Stock Stain 1%, Aqueous 100 ml. Newcomer Supply Eosin-Phloxine Stain Set solutions are aqueous based and intended to give an extra fine touch to routine hematoxylin and eosin (H&E) stains and can be used in either manual or automated staining platforms. Differentiation of muscle, connective tissue and epithelial elements tend to be sharper and with better demonstration than with the traditional eosin y solution alone. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0068658 Supplier Newcomer Supply Supplier No. 1082A
Solution A: Eosin Y Stock Stain 1%, Aqueous 1000 ml, Solution B: Phloxine B Stock Stain 1%, Aqueous 100 ml. Newcomer Supply Eosin-Phloxine Stain Set solutions are aqueous based and intended to give an extra fine touch to routine hematoxylin and eosin (H&E) stains and can be used in either manual or automated staining platforms. Differentiation of muscle, connective tissue and epithelial elements tend to be sharper and with better demonstration than with the traditional eosin y solution alone. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.