Learn More
Newcomer Supply Eosin Y Working Solution, 1 gallon. Ready to use counterstain in the routine hematoxylin and eosin stain and can be used in either manual or automated staining platforms.
SDP
Catalog No. NC0916717
Encompass
Use: Counterstain for H&E Stain. Newcomer Supply Eosin Y Working Solution is a ready to use counterstain in the routine hematoxylin and eosin stain and can be used in either manual or automated staining platforms. Eosin’s particular value is in its ability to distinguish between the cytoplasm of different types of cells by staining cytoplasmic components differing shades and intensities of pink to red. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0916717 Supplier Newcomer Supply Supplier No. 1072C
Use: Counterstain for H&E Stain. Newcomer Supply Eosin Y Working Solution is a ready to use counterstain in the routine hematoxylin and eosin stain and can be used in either manual or automated staining platforms. Eosin’s particular value is in its ability to distinguish between the cytoplasm of different types of cells by staining cytoplasmic components differing shades and intensities of pink to red. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.