Learn More
Polysciences, Inc. Eosin Y, 0.5% Alcoholic Solution, Used as Counterstain, Contains Acetic Acid 1000ml
SDP
Catalog No. NC9022609
Encompass
Eosin Y, 0.5% Alcoholic Solution, Used as Counterstain, Contains Acetic Acid, Store at 25 deg.C, Red Colored Liquid Alcohol Odor, Vapor Pressure: 44.6mmHg, BP: 173, MP: -148, Specific Gravity: 0.86, Store at 25 deg.C
Catalog No. NC9022609 Supplier Polysciences, Inc. Supplier No. 098591000
Eosin Y, 0.5% Alcoholic Solution, Used as Counterstain, Contains Acetic Acid, Store at 25 deg.C, Red Colored Liquid Alcohol Odor, Vapor Pressure: 44.6mmHg, BP: 173, MP: -148, Specific Gravity: 0.86, Store at 25 deg.C