Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Eosin Y, 1% Aqueous Solution

Catalog No. ES8901
Change view
Click to view available options
Quantity:
1 x 950 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8901 1 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES8901 Supplier ENG Scientific Supplier No. 8901
Add to Cart
Add to Cart

Eosin Y, 1% Aqueous Solution is used as background contrast to nuclear stains. Depicts cytoplasmic detail of the cell and tissue structure.

  • Solutions are used mainly as background contrast to nuclear stains, i.e., hematoxylin-eosin sequence.
  • Eosin Y stain depicts the cytoplasmic detail of the cell and the tissue structure.
  • Contents: Eosin Y, Ethanol (where applicable)
  • These solutions are made from certified dyes.

Specifications

Chemical Name or Material Eosin Y
CAS 15086-94-9
Packaging Bottle
Quantity 1 x 950 mL

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.