Learn More
Medchemexpress LLC Eosin Y | 15086-94-9 | 99.47% | 25 G
SDP
Catalog No. 5000413896
Encompass_Preferred
Eosin Y (Acid Red 87 free base) is a soluble acid red dye molecule with wide application in organic synthesis as a photoredox catalyst. It is also used in cell staining, as a pH indicator, an indicator in analytical halide determination by Fajans, and as a dye pigment. It is the most widely used counterstain in routine staining of histological tissue sections (erythrocytes, collagen, epithelial cells, etc.).
  • Soluble acid red dye molecule
  • Applicable as a photoredox catalyst in organic synthesis
  • Used in cell staining, as a pH indicator, and in analytical halide determination
  • Commonly used counterstain in histological tissue sections
  • Versatile for various applications including organic synthesis and biological staining
  • Effective as a photoredox catalyst
  • Reliable for routine staining of histological tissue sections
Catalog No. 50-004-13896 Supplier Medchemexpress LLC Supplier No. 25GHYD0505
Eosin Y (Acid Red 87 free base) is a soluble acid red dye molecule with wide application in organic synthesis as a photoredox catalyst. It is also used in cell staining, as a pH indicator, an indicator in analytical halide determination by Fajans, and as a dye pigment. It is the most widely used counterstain in routine staining of histological tissue sections (erythrocytes, collagen, epithelial cells, etc.).
  • Soluble acid red dye molecule
  • Applicable as a photoredox catalyst in organic synthesis
  • Used in cell staining, as a pH indicator, and in analytical halide determination
  • Commonly used counterstain in histological tissue sections
  • Versatile for various applications including organic synthesis and biological staining
  • Effective as a photoredox catalyst
  • Reliable for routine staining of histological tissue sections