Learn More
Medchemexpress LLC Eosin Y | 15086-94-9 | MFCD00036189 | 99.4% | 5 G
SDP
Catalog No. 5000410243
Encompass_Preferred
Eosin Y (Acid Red 87 free base) is a soluble acid red dye molecule with wide application in organic synthesis as a photoredox catalyst. It is also used in cell staining, as a pH indicator, as an indicator in the analytical halide determination by Fajans, and as a dye pigment (e.g., in lipsticks). It is the most widely used counterstain in routine staining of histological tissue sections, including erythrocytes, collagen, and epithelial cells.
  • Soluble acid red dye molecule
  • Applicable as a photoredox catalyst in organic synthesis
  • Used for cell staining
  • Functions as a pH indicator
  • Used as an indicator in analytical halide determination by Fajans
  • Can be used as a dye pigment
  • Most widely used counterstain in routine histological tissue section staining
Catalog No. 50-004-10243 Supplier Medchemexpress LLC Supplier No. HYD05055G
Eosin Y (Acid Red 87 free base) is a soluble acid red dye molecule with wide application in organic synthesis as a photoredox catalyst. It is also used in cell staining, as a pH indicator, as an indicator in the analytical halide determination by Fajans, and as a dye pigment (e.g., in lipsticks). It is the most widely used counterstain in routine staining of histological tissue sections, including erythrocytes, collagen, and epithelial cells.
  • Soluble acid red dye molecule
  • Applicable as a photoredox catalyst in organic synthesis
  • Used for cell staining
  • Functions as a pH indicator
  • Used as an indicator in analytical halide determination by Fajans
  • Can be used as a dye pigment
  • Most widely used counterstain in routine histological tissue section staining