Learn More
Ethos Biosciences Eosin Y 1PCT Alcoholic 16oz
Catalog No. 5000819195
Encompass_Preferred
Eosin Y 1% Alcoholic is a ready-to-use cytoplasmic counterstain formulated for routine histology and cytology applications Commonly used in hematoxylin and eosin (H E) staining procedures it provides clear pink to red coloration of cytoplasmic components connective tissue and extracellular structures enhancing contrast with nuclear stains The alcoholic formulation supports rapid penetration and consistent staining intensity making it suitable for manual and automated staining systems Designed for use in clinical research and educational laboratories Eosin Y 1% Alcoholic delivers reliable reproducible results for routine tissue evaluation
Catalog No. 50-008-19195 Supplier Ethos Biosciences Supplier No. 700616
Eosin Y 1% Alcoholic is a ready-to-use cytoplasmic counterstain formulated for routine histology and cytology applications Commonly used in hematoxylin and eosin (H E) staining procedures it provides clear pink to red coloration of cytoplasmic components connective tissue and extracellular structures enhancing contrast with nuclear stains The alcoholic formulation supports rapid penetration and consistent staining intensity making it suitable for manual and automated staining systems Designed for use in clinical research and educational laboratories Eosin Y 1% Alcoholic delivers reliable reproducible results for routine tissue evaluation