Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Eosin-Y Alcoholic

Catalog No. 23730609 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-730-609 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-730-609 Supplier Scigen Supplier No. 7115
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An alcoholic eosin that allows for exceptional differentiation and contrast between cytoplas-mic components and nuclei.

Scigen™ Eosin-Y is an alcoholic eosin that allows for exceptional differentiation and contrast between cytoplas-mic components and nuclei. Scigen™ Eosin-Y stains cytoplasm three shades of pink and red. Prior to staining in Eosin-Y tissue slides should be rinsed in alcohol. The concentration of the alcohol rinse can range from 70% to 95%.

Staining time in Scigen™ Eosin Y depends on the use and personal preference; the following are guidelines that help establish a custom protocol for your specific needs. When staining routine hematoxylin and eosin sections, good results can be obtained with Phantom Eosin Y with a staining time of 0.5 to 1.5 minutes. A staining time of 0.5 minutes is recommended as an initial staining time, with an increase of 0.5 minute intervals until desired color and density of the cytoplasm staining is achieved.

Specifications

Certifications/Compliance Traceable Certififed Biological Stain
Color Eosin-Y
Description Provides excellent contrast between cytoplasmic components and the nuclei of cells
For Use With (Application) 3 Color Contrasting Stain
Includes Staining Protocol
Material Alochol Based
Packaging Light Resistant Bottle
Quantity 4/Cs.
Solution Type Alcohol-Aqueous Base
Storage Requirements Room Temperature, Dark Storage
Synonym Eosin-Y
Product Type Alcoholic Eosin Y Stain
Show More Show Less

FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.