Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Eosin Y, Certified, C.I. DcE-40 10 GR
SDP

Catalog No. 50980444
Encompass_Preferred

(Acid Red 87, Eosin Yellowish)
CAS #17372-87-1 C₂cH₆Br₄Na₂O₅ F.W. 691.88
Solubility: 44.0% Water; 2.0% Alcohol; 25.0% Cellosolve; 27.5% Glycol; 0.0% Xylene

Eosin Y is used with Hematoxylin for showing cell nuclei in embryos and kidneys (Conn, "Biological Stain" 9th ed., The Williams and Wilkens Co., Baltimore 1977), as a component of Wright's stain for blood corpuscles (Wright, J. Med. Res., 7, 138 (1902), and for staining bone marrow to show cell morphology (Bech, "Lab. Manual of Hematological Tech.", W. B. Saunders Co., Phila., 1908, p. 233).

Catalog No. 50-980-444 Supplier Electron Microscopy Sciences Supplier No. 14850
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Acid Red 87, Eosin Yellowish)
CAS #17372-87-1 C₂cH₆Br₄Na₂O₅ F.W. 691.88
Solubility: 44.0% Water; 2.0% Alcohol; 25.0% Cellosolve; 27.5% Glycol; 0.0% Xylene

Eosin Y is used with Hematoxylin for showing cell nuclei in embryos and kidneys (Conn, "Biological Stain" 9th ed., The Williams and Wilkens Co., Baltimore 1977), as a component of Wright's stain for blood corpuscles (Wright, J. Med. Res., 7, 138 (1902), and for staining bone marrow to show cell morphology (Bech, "Lab. Manual of Hematological Tech.", W. B. Saunders Co., Phila., 1908, p. 233).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.