Learn More
Selleck Chemical LLC Eosin Y Disodium
SDP
Catalog No. 5000003581
Encompass_Preferred
Eosin Y is a form of eosin It is most commonly used as an acidic red stain for highlighting cytoplasm material in samples
Catalog No. 50-000-03581 Supplier Selleck Chemical LLC Supplier No. S552125MG
Eosin Y is a form of eosin It is most commonly used as an acidic red stain for highlighting cytoplasm material in samples