Learn More
Eosin Y Disodium Salt, Sigma-Aldrich
Catalog No. 01258811 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
01-258-811 100 g
1 options
Catalog No. 01-258-811 Supplier MilliporeSigma Supelco Supplier No. E4382100G
Add to Cart
Add to Cart
Biological Stain Commission-certified xanthene dye for differential staining of connective tissue and cytoplasm.
Eosin Y is a xanthene dye and is used for the differential staining of connective tissue and cytoplasm. In histopathology, it is applied as a counterstain after hematoxylin and before methylene blue. It is also used as a background stain, thereby giving contrast to the nuclear stains.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Type Eosin Y