Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution I - Eosin Y

Catalog No. ES6921
Change view
Click to view available options
Quantity:
1 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6921 1 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES6921 Supplier ENG Scientific Supplier No. 6921
Add to Cart
Add to Cart
This item is not returnable. View return policy

Solution I - Eosin Y is used in staining eosinophils from secretions of the nasal passage, sinuses, bronchi and urinary sediment.

  • For Eosinophil Stain Kit.

Specifications

Chemical Name or Material Eosin Y
CAS 15086-94-9
Packaging Bottle
Quantity 1 x 250 mL

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.