Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Free Acid, MP Biomedicals

Catalog No. ICN15792205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15792205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15792205 Supplier MP Biomedicals, Inc Supplier No. 0215792205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15086-94-9
Molecular Formula C20H8Br4O5
Synonym 2,4,5,7-Tetrabromofluorescein, Eosin yellowish

Specifications

Assay Percent Range ∽ 99%
Quantity 5 g
Formula Weight 647.9
Chemical Name or Material Eosin Y Free Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.