Learn More
Azer Scientific Eosin-Y (Intensified), iCON™ Staining System
SDP
Catalog No. 21377133
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
4 x 16 oz.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-133 16 oz.
21-377-134 1 gal.
21-377-135 4 x 1 gal.
21-377-136 4 x 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-377-133 Supplier Azer Scientific Supplier No. ES3653116
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Produces a wide range of color intensities, staining brilliance with exception of its pH balanced technology.
  • Designed to produce consistent quality results
  • May be used on all automatic stainers as well as in manual staining
  • Produces a wide range of color intensities and staining brilliance
  • Alcoholic Intensified Eosin-Y has the attributes of traditional Eosin-Y stains with the exception of its pH balanced technology
  • Intensified eosin-Y features a pH range of 4.6 to 5 results in eosin staining, three shades of pink to red and resolves the problem of inconsistent eosin staining
  • When the pH of eosin is below pH 4 it binds nonspecifically to proteins causing the stained cytoplasm to appear muddy
Specifications
Type Stain Kit
Quantity 16 oz.