Learn More
Azer Scientific Eosin-Y Phloxine-B, iCON™ Staining System
SDP
Catalog No. 21377140
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
4 x 16 oz.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-140 4 x 16 oz.
21-377-137 16 oz.
21-377-138 1 gal.
21-377-139 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-377-140 Supplier Azer Scientific Supplier No. ES365334X16
Add to Cart
Add to Cart
This item is not returnable. View return policy
Producing crisp and well-defined detail is our proprietary family of pH-balanced
  • Producing crisp and well-defined detail is our proprietary family of pH-balanced iCON™ Intensified Cytoplasmic Stains
  • This family of cytoplasmic stains consists of two distinct products designed to produce consistent quality results
  • Designed to produce consistent quality results
  • May be used on all automatic stainers as well as in manual staining
  • Produces a wide range of color intensities and staining brilliance
Specifications
For Use With (Application) Automatic Stainers
Quantity 4 x 16 oz.
Product Type Stain Kit