Learn More
StatLab™ Eosin-Y Phloxine B Stain
GSA_VASDP
Catalog No. STLSL104
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
STLSL104 4 x 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL104 Supplier StatLab™ Supplier No. SL104
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Cytoplasmic counterstain that provides a brighter reddish stain in muscle and other connective tissue.
  • Provides a more dramatic color contrast than traditional Eosin-Y
  • StatLab™'s Vintage Stain Line features more common hybrid stains
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 500 mL
Product Type Eosin Stain