Learn More
Bioworld Eosin Y, Sodium Salt, 100 g
Catalog No. 50199207
Encompass_Preferred
Specifications:Certified GradeA histological counterstain against hematoxylin. Dye Content (%) : 88Lambda Max (nm) : 515 - 518
Catalog No. 50-199-207 Supplier Bioworld Supplier No. 217700151
Specifications:Certified GradeA histological counterstain against hematoxylin. Dye Content (%) : 88Lambda Max (nm) : 515 - 518