Learn More
Scytek Laboratories Eosin Y Solution, 500 mL, Newly Formulated
SDP
Catalog No. NC1822444
Encompass_Preferred
Eosin Y Solution (Modified Alcoholic) is intended for use in histology and cytology applications. This newly formulated Eosin provides the benefits of a traditional alcoholic formula with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, reduced tendency to spill over container, hands, and countertops, and improved surface tension to remain on tissue section.
This item is not returnable. View return policy
Catalog No. NC1822444 Supplier Scytek Laboratories Supplier No. EYB500
Eosin Y Solution (Modified Alcoholic) is intended for use in histology and cytology applications. This newly formulated Eosin provides the benefits of a traditional alcoholic formula with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, reduced tendency to spill over container, hands, and countertops, and improved surface tension to remain on tissue section.