Learn More
Newcomer Supply Eosin Y Stock Stain 1%, Alcoholic, 1 gallon
SDP
Catalog No. NC9801930
Encompass
Use: Counterstain for H&E Stain. Newcomer Supply Eosin Y Stock Stain 1%, Alcoholic provides the key component of an alcohol based Eosin Y Working Solution in the routine hematoxylin and eosin stain and can be used in either manual or automated staining platforms.  Eosin’s particular value is in its ability to distinguish between the cytoplasm of different types of cells by staining cytoplasmic components differing shades and intensities of pink to red. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9801930 Supplier Newcomer Supply Supplier No. 1070D
Use: Counterstain for H&E Stain. Newcomer Supply Eosin Y Stock Stain 1%, Alcoholic provides the key component of an alcohol based Eosin Y Working Solution in the routine hematoxylin and eosin stain and can be used in either manual or automated staining platforms.  Eosin’s particular value is in its ability to distinguish between the cytoplasm of different types of cells by staining cytoplasmic components differing shades and intensities of pink to red. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.