Learn More
Electron Microscopy Sciences Eosin Y Stock Solution 1% Aqueous 500 ML
SDP
Catalog No. 5031909
Encompass_Preferred
Eosin Y Stock Solution in a 1% aqueous solution for Eosin-Phloxine Counterstain.
Catalog No. 50-319-09 Supplier Electron Microscopy Sciences Supplier No. 2676301
Eosin Y Stock Solution in a 1% aqueous solution for Eosin-Phloxine Counterstain.