Learn More
Cayman Chemical EosIn YsodIum salt 5g
SDP
Catalog No. 502912351
Encompass_Preferred
A dye and brominated fluorescein derivative; exists as a tautomer in aqueous solutions; has commonly been used in H&E staining to stain the extracellular matrix and cytoplasmic proteins; ex/em = 526/544 nm, respectively; has been used as a photoredox catalyst in various organic synthesis reactions; a non-selective inhibitor of histone methyltransferases at 100 µM
Catalog No. 50-291-2351 Supplier Cayman Chemical Supplier No. 363355 G
A dye and brominated fluorescein derivative; exists as a tautomer in aqueous solutions; has commonly been used in H&E staining to stain the extracellular matrix and cytoplasmic proteins; ex/em = 526/544 nm, respectively; has been used as a photoredox catalyst in various organic synthesis reactions; a non-selective inhibitor of histone methyltransferases at 100 µM