Learn More
Euthanex The 512-A1-DS is a non-compensated single CO2 flow meter with flow rate range of 0.5 to 12 LPM.
Catalog No. NC1286486
Encompass
The 512-A1-DS non-compensated CO2 single flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. Connects directly to point of use CO2 source with a standard male DISS 1080 fitting. We also offer male DISS fitting in 1/8"""" and 1/4"""" NPT. The DISS includes an output line with quick disconnect for easy connection to our lids or induction chambers.
Catalog No. NC1286486 Supplier Euthanex Supplier No. 512A1DS
The 512-A1-DS non-compensated CO2 single flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. Connects directly to point of use CO2 source with a standard male DISS 1080 fitting. We also offer male DISS fitting in 1/8"""" and 1/4"""" NPT. The DISS includes an output line with quick disconnect for easy connection to our lids or induction chambers.