Learn More
Biotium Ep-CAM / CD326 (Rat)(GZ-20), CF594 conjugate, 0.1mg/mL
Supplier:  Biotium BNC940382500
Encompass_Preferred
Epitope of GZ-1 and GZ-20 monoclonal antibodies are different. EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on basolateral cell surface in most simple epithelia and a vast majority of carcinomas. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-252-3146