Learn More
Cayman Chemical EP1 Receptr Blockng PeptID 1ea
SDP
Catalog No. 502978728
Encompass_Preferred
An internal standard for the quantification of almorexant by GC- or LC-MS
Catalog No. 50-297-8728 Supplier Cayman Chemical Supplier No. 3017401 EA
An internal standard for the quantification of almorexant by GC- or LC-MS