Learn More
Cayman Chemical EP2 Receptor Blocking Peptide, 1 ea
SDP
Catalog No. 502978729
Encompass_Preferred
Peptide Sequence: human EP2 receptor amino acids 335-358 (SLRTQDATQTSCSTQSDASKQADL){3178} · To be used in conjunction with Cayman’s EP2 receptor polyclonal antibody (Catalog No. 101750) to block protein-antibody complex formation during immunochemical anaylsis for the EP2 receptor.
Catalog No. 50-297-8729 Supplier Cayman Chemical Supplier No. 3017501 EA
Peptide Sequence: human EP2 receptor amino acids 335-358 (SLRTQDATQTSCSTQSDASKQADL){3178} · To be used in conjunction with Cayman’s EP2 receptor polyclonal antibody (Catalog No. 101750) to block protein-antibody complex formation during immunochemical anaylsis for the EP2 receptor.