Learn More
Cayman Chemical EP3 Receptor Blocking Peptide, 1 ea
SDP
Catalog No. 502978730
Encompass_Preferred
Peptide Sequence: Human EP3 receptor sequence amino acids 308-327 (NQTSVEHCKTHTEKQKECNF){3180,1900} • To be used in conjunction with Cayman’s EP3 Receptor Polyclonal Antibody (Item No. 101760) to block protein-antibody complex formation during immunochemical analysis for the EP3 receptor.
Catalog No. 50-297-8730 Supplier Cayman Chemical Supplier No. 3017601 EA
Peptide Sequence: Human EP3 receptor sequence amino acids 308-327 (NQTSVEHCKTHTEKQKECNF){3180,1900} • To be used in conjunction with Cayman’s EP3 Receptor Polyclonal Antibody (Item No. 101760) to block protein-antibody complex formation during immunochemical analysis for the EP3 receptor.