Learn More
ABclonal Technology EP400 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1215120UL
Encompass_Preferred
Predicted to enable several functions, including ATP binding activity, ATP-dependent chromatin remodeler activity, and protein antigen binding activity. Involved in histone H2A acetylation and histone H4 acetylation. Part of NuA4 histone acetyltransferase complex and Swr1 complex.
Catalog No. 50-001-14606