Learn More
Hach Company Xenosep SPE Starter Kit with Solvent Recovery, EPA Method 1664A
Catalog No. 501996867 Shop All Hach Company Products
Encompass
Xenosep Solid-Phase Extraction (SPE) Systems offer the easiest and most cost effective way to test for Oil and Grease using EPA Method 1664A. Fast, in-house results enable you to maintain control over your plant discharges in real time without the complications of the hexane LLE procedure. Xenosep SPE filters feature a unique triple layer that prevents premature clogging and flows 10 times faster than typical SPE filters.It is now possible to easily achieve reliable, consistent Oil and Grease results and decrease your sample turnaround time. A free training video on CD is included with each Starter Kit to show your lab how to perform EPA Method 1664A by SPE.Xenosep Starter Kit with solvent recovery is the same as the Xenosep Starter Kit without Solvent Recovery except it includes a distillation assembly and a total of 7 flat-sided boiling flasks.
Catalog No. 50-199-6867 Supplier Hach Company Supplier No. 2943200
Xenosep Solid-Phase Extraction (SPE) Systems offer the easiest and most cost effective way to test for Oil and Grease using EPA Method 1664A. Fast, in-house results enable you to maintain control over your plant discharges in real time without the complications of the hexane LLE procedure. Xenosep SPE filters feature a unique triple layer that prevents premature clogging and flows 10 times faster than typical SPE filters.It is now possible to easily achieve reliable, consistent Oil and Grease results and decrease your sample turnaround time. A free training video on CD is included with each Starter Kit to show your lab how to perform EPA Method 1664A by SPE.Xenosep Starter Kit with solvent recovery is the same as the Xenosep Starter Kit without Solvent Recovery except it includes a distillation assembly and a total of 7 flat-sided boiling flasks.