Learn More
Hach Company Xenosep EPA 1664A Standard, 1x10 mL, 4mg/mL
Supplier: Hach Company 2943702
Xenosep EPA 1664A Standard, 1x10 mL, 4mg/mL. Pipet 10 mL to achieve the 40mg PAR (Precision and Recovery) Standard needed for the method. Each bottle contains enough standard to QC one batch of samples. 10ml.