Learn More
Crescent Chemical Co Inc EPA 200.8 SOLN B 2COMP 100ML
SDP
Catalog No. 5068562
Encompass_Preferred
Calibration Standards for Method 200.8 Standard Solution B for 200.8 2 Components in 1% HNO3 Ag 10 UgmL Ba 10 UgmL CAS Size - 100 ml Catalog Number - CC5605.1
Catalog No. 50-685-62 Supplier Crescent Chemical Co Inc Supplier No. CTCC5605.1
Calibration Standards for Method 200.8 Standard Solution B for 200.8 2 Components in 1% HNO3 Ag 10 UgmL Ba 10 UgmL CAS Size - 100 ml Catalog Number - CC5605.1