Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA 501/601 Trihalomethanes Calibration Mix, Certified Reference Material, 2,000 μg/mL, MilliporeSigma™ Supelco™

Catalog No. 111017173 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7173 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7173 Supplier MilliporeSigma Supelco Supplier No. 4M8140U
Add to Cart
Add to Cart

2,000 μg/mL Each Component in Methanol

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 2,000 μg/mL each component in methanol
Type Organic Pollutant Standard
Quantity 5 x 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Bromodichloromethane; Bromoform; Chloroform; Dibromochloromethane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.