Learn More
Crescent Chemical Co Inc EPA 502.2 VOC GAS 6COMP 1ML
SDP
Catalog No. 5067799
Encompass_Preferred
CLP Volatiles Calibration Mix #2 - Gases, 6 Components Bromomethane, Chloroethane, Chloromethane, Dichlorodifluoromethane, Trichlorofluoromethane, Vinyl chloride CAS Size - 1mL Catalog Number - CC2006.10
Catalog No. 50-677-99 Supplier Crescent Chemical Co Inc Supplier No. CTCC2006.10
CLP Volatiles Calibration Mix #2 - Gases, 6 Components Bromomethane, Chloroethane, Chloromethane, Dichlorodifluoromethane, Trichlorofluoromethane, Vinyl chloride CAS Size - 1mL Catalog Number - CC2006.10