Learn More
Crescent Chemical Co Inc EPA 515.1 ACIDS 16COMP 1ML
SDP
Catalog No. 5067853
Encompass_Preferred
EPA Method 515.1-Acids Acifluorfen-100 ugmL, Bentazone-200 ugmL, Chloramben-100 ugmL, 2,4-D-200 ugmL, Dalapon1,300 ugmL, 2,4-DB-800ugmL, DCPA diacid-100 ugmL, Dicamba-100 ugmL, 3,5-Dichlorobenzoic acid-100 ugmL, Dichloroprop-300 ugmL, Dinoseb-200 ugm
Catalog No. 50-678-53 Supplier Crescent Chemical Co Inc Supplier No. CTCC2058
EPA Method 515.1-Acids Acifluorfen-100 ugmL, Bentazone-200 ugmL, Chloramben-100 ugmL, 2,4-D-200 ugmL, Dalapon1,300 ugmL, 2,4-DB-800ugmL, DCPA diacid-100 ugmL, Dicamba-100 ugmL, 3,5-Dichlorobenzoic acid-100 ugmL, Dichloroprop-300 ugmL, Dinoseb-200 ugm